Ethyl pyruvate ≥98%
Supplier: BeanTown Chemical
Synonyms:
Ethyl 2-oxopropanoate, 2-Oxopropionic acid ethyl ester, α-Ketopropionic acid ethyl ester, Pyruvic acid ethyl ester, 2-Ketopropionic acid ethyl ester
211775-250G
211775-500G
211775-50G
BT211775-250GEA
157.56
USD
BT211775-250G
BT211775-500G
BT211775-50G
Ethyl pyruvate ≥98%
Ethyl pyruvate
CAS: 617-35-6; EC No: 210-511-2; MDL No: MFCD00009123
UN No: UN3272; Haz Class: 3; Packing Group: III
Liquid; Linear Formula: CH2COCOOCH2CH3; Molecular Formula: C5H8O3; MW: 116.12
Boiling Point: 144°; Flash point: 46°C (115°F)
Density (g/mL): 1.045; Refractive Index: 1.404
- Refractive index: 1.404
- Liquid
Formula:
C₅H₈O₃ Boiling Pt: 144 °C Density: 1.045 g/mL Flash Pt: 46 °C (115 °F) |
MDL Number:
MFCD00009123 CAS Number: 617-35-6 EINECS: 210-511-2 UN: 3272 ADR: 3,III |
Specification Test Results
Assay | 98% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...