2-Chloro-4,5-difluorobenzonitrile ≥97%

Supplier: BeanTown Chemical

211715-10G 211715-2G
BT211715-10GEA 202 USD
BT211715-10G BT211715-2G
2-Chloro-4,5-difluorobenzonitrile ≥97%
2-Chloro-4,5-difluorobenzonitrile

CAS: 135748-34-4; MDL No: MFCD02871619 UN No: UN3439; Haz Class: 6.1; Packing Group: III Solid; Molecular Formula: C7H2ClF2N; MW: 173.55 Melting Point: 35-40°


  • Solid

Formula: C₇H₂ClF₂N
Melting Pt: 35…40 °C
MDL Number: MFCD02871619
CAS Number: 135748-34-4
UN: 3439
ADR: 6.1,III

Order Now

Specification Test Results

Assay 97%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR