(±)-2-Butanol ≥99%

Supplier: BeanTown Chemical

Synonyms: (±)-sec-Butanol, (±)-sec-Butyl alcohol, (±)-Ethyl methyl carbinol, (±)-2-Hydroxy butane, sec-Butanol, sec-Butyl alcohol, (±)-Methyl ethyl carbinol

211685-2.5L 211685-500ML
BT211685-2.5LEA 85.09 USD
BT211685-2.5L BT211685-500ML
(±)-2-Butanol ≥99%
(±)-2-Butanol

CAS: 78-92-2; EC No: 201-158-5; MDL No: MFCD00004569; RTECS: EO1750000 UN No: UN1120; Haz Class: 3; Packing Group: III Liquid; Linear Formula: CH3CH2CH(OH)CH3; Molecular Formula: C4H10O; MW: 74.12 Melting Point: -115°; Boiling Point: 98°; Flash point: 27°C (81°F) Density (g/mL): 0.808; Refractive Index: 1.414


  • RTECS: EO1750000
  • Refractive index: 1.414
  • Liquid

Formula: H₃CCH₂CH(OH)CH₃
MW: 74.12 g/mol
Boiling Pt: 98 °C
Melting Pt: –115 °C
Density: 0.808 g/mL
Flash Pt: 27 °C (81 °F)
MDL Number: MFCD00004569
CAS Number: 78-92-2
EINECS: 201-158-5
UN: 1120
ADR: 3,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=1dea3607167631fbfcf7751c20b71f345ad24c19&vid=010e04e8f508fd3d501873d20c2349817834b6e8&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay 99%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR