Indium(III) isopropoxide ≥99.9% (trace metals basis)

Supplier: BeanTown Chemical
Danger

211680-10G 211680-2G 211680-500MG
BT211680-10GEA 862 USD
BT211680-10G BT211680-2G BT211680-500MG
Indium(III) isopropoxide ≥99.9% (trace metals basis)
Indium (III) isopropoxide

CAS: 38218-24-5; MDL No: MFCD00210318 UN No: UN3181; Haz Class: 4.1; Packing Group: III Powder; Linear Formula: In(OCH(CH3)2)3; MW: 292.08 Boiling Point: 83-87°; Flash point: 12°C (54°F) Density (g/mL): 0.808 Moisture Sensitive

Formula: C₉H₂₁InO₃
MW: 292.08 g/mol
Boiling Pt: 83…87 °C
Density: 0.808
Flash Pt: 12 °C
MDL Number: MFCD00210318
CAS Number: 38218-24-5
UN: 2810
ADR: 4.1 III

Order Now

Specification Test Results

Assay 99.9%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR