O-Phenylenediamine ≥98%
Supplier: BeanTown Chemical
Synonyms:
1,2-Phenylendiamine, 1,2-Phenylenediamine, 1,2-Diaminobenzene, OPD
211675-1KG
211675-250G
211675-25G
BT211675-1KGEA
182.66
USD
BT211675-1KG
BT211675-250G
BT211675-25G
O-Phenylenediamine ≥98%
O-Phenylenediamine
CAS: 95-54-5; EC No: 202-430-6; MDL No: MFCD00007721; RTECS: SS7875000
UN No: UN1673; Haz Class: 6.1; Packing Group: III
Solid; Linear Formula: C6H4(NH2)2; Molecular Formula: C6H8N2; MW: 108.14
Melting Point: 98-102°; Boiling Point: 256-258°; Flash point: 136°C (277°F)
Density (g/mL): 1.03
Moisture Sensitive
- RTECS: SS7875000
- Moisture Sensitive
- Solid
Formula:
C₆H₈N₂ MW: 108.14 g/mol Boiling Pt: 256…258 °C Melting Pt: 98…102 °C Density: 1.03 g/mL Flash Pt: 136 °C (277 °F) |
MDL Number:
MFCD00007721 CAS Number: 95-54-5 EINECS: 202-430-6 UN: 1673 ADR: 6.1,III |
Specification Test Results
Assay | 98% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...