(±)-4-Vinylcyclohexene
Supplier: BeanTown Chemical
Synonyms:
(±)-4-Ethenyl-1-cyclohexene, 4-Vinyl-1-cyclohexene
211670-25ML
211670-5ML
BT211670-25MLEA
171
USD
BT211670-25ML
BT211670-5ML
(±)-4-Vinylcyclohexene
(±)-4-Vinylcyclohexene
CAS: 100-40-3; EC No: 202-848-9; MDL No: MFCD00001576; RTECS: GW6650000
UN No: UN1993; Haz Class: 3; Packing Group: II
Liquid; Linear Formula: C6H9CH=CH2; Molecular Formula: C8H12; MW: 108.18
Melting Point: -101°; Boiling Point: 126-127°; Flash point: 16°C (61°F)
Density (g/mL): 0.832; Refractive Index: 1.463
Air Sensitive
- RTECS: GW6650000
- Refractive index: 1.463
- Air Sensitive
- Liquid
Formula:
C₈H₁₂ MW: 108.18 g/mol Boiling Pt: 126…127 °C Melting Pt: –101 °C Density: 0.832 g/mL Flash Pt: 16 °C (61 °F) |
MDL Number:
MFCD00001576 CAS Number: 100-40-3 EINECS: 202-848-9 UN: 1993 ADR: 3,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...