N,N-Dimethylformamide dimethyl acetal ≥97%

Supplier: BeanTown Chemical

Synonyms: 1,1-Dimethoxytrimethylamine, Esterate M, N,N-Dimethylformamide Dimethyl Acetal, 1,1-Dimethoxy-N,N-dimethylmethylamine, DMF-DMA

211640-100G 211640-25G 211640-500G
BT211640-100GEA 135.44 USD
BT211640-100G BT211640-25G BT211640-500G
N,N-Dimethylformamide dimethyl acetal ≥97%
N,N-Dimethylformamide dimethyl acetal

CAS: 4637-24-5; EC No: 225-063-3; MDL No: MFCD00008482 UN No: UN1993; Haz Class: 3; Packing Group: II Liquid; Linear Formula: (CH3)2NCH(OCH3)2; Molecular Formula: C5H13NO2; MW: 119.16 Melting Point: -85°; Boiling Point: 102-103°/720 mmHg; Flash point: 5°C (41°F) Density (g/mL): 0.897; Refractive Index: 1.396 Moisture Sensitive


  • Refractive index: 1.396
  • Moisture Sensitive
  • Liquid

Formula: C₅H₁₃NO₂
MW: 119.16 g/mol
Boiling Pt: 102…103 °C/720 mmHg
Melting Pt: –85 °C
Density: 0.897 g/mL
Flash Pt: 5 °C (41 °F)
MDL Number: MFCD00008482
CAS Number: 4637-24-5
EINECS: 225-063-3
ADR: 3,II

Order Now

Specification Test Results

Assay 97%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR