Nickel (II) sulfate hexahydrate ≥98%
Supplier: BeanTown Chemical
211535-2KG
211535-500G
BT211535-2KGEA
213.34
USD
BT211535-2KG
BT211535-500G
Nickel (II) sulfate hexahydrate ≥98%
Nickel(II) sulphate hexahydrate
CAS: 10101-97-0; EC No: 232-104-9; MDL No: MFCD00149813; RTECS: QR9600000
UN No: UN3288; Haz Class: 6.1; Packing Group: III
Crystalline/Powder; Linear Formula: NiSO4·6H2O; MW: 262.86
Density (g/mL): 2.070
- RTECS: QR9600000
- Crystalline/Powder
Formula:
NiSO₄·6H₂O Density: 2.070 g/mL |
MDL Number:
MFCD00149813 CAS Number: 10101-97-0 EINECS: 232-104-9 UN: 3288 ADR: 6.1,III |
Specification Test Results
Assay | 98% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...