Disodium hexachloroplatinate hexahydrate
Supplier: BeanTown Chemical
Synonyms:
Sodium hexachloroplatinate(IV) hexahydrate, Sodium hexachloroplatinate (IV) hexahydrate
211530-1G
211530-5G
BT211530-1GEA
121.12
USD
BT211530-1G
BT211530-5G
Disodium hexachloroplatinate hexahydrate
Disodium hexachloroplatinate hexahydrate
CAS: 19583-77-8; EC No: 240-983-5; MDL No: MFCD00149172
UN No: UN3288; Haz Class: 6.1; Packing Group: III
Crystalline; Linear Formula: Na2PtCl6·6H2O; MW: 561.88
Density (g/mL): 2.5
- Crystalline
Formula:
Cl₆Na₂Pt·6H₂O Density: 2.5 g/mL |
MDL Number:
MFCD00149172 CAS Number: 19583-77-8 EINECS: 240-983-5 UN: 3288 ADR: 6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...