5-Sulfosalicylic acid dihydrate ≥98%
Supplier: BeanTown Chemical
Synonyms:
2-Hydroxy-5-sulphobenzoic acid dihydrate
211520-1KG
211520-250G
BT211520-1KGEA
120
USD
BT211520-1KG
BT211520-250G
5-Sulfosalicylic acid dihydrate ≥98%
5-Sulphosalicylic acid dihydrate
CAS: 5965-83-3; EC No: 202-555-6; MDL No: MFCD00007508; RTECS: VO6500000
UN No: UN2585; Haz Class: 8; Packing Group: III
Crystalline; Molecular Formula: C7H6O6S·2H2O; MW: 254.21
Melting Point: 105-110°
Light Sensitive
- RTECS: VO6500000
- Light Sensitive
- Crystalline
Formula:
HO₃SC₆H₃-2-(OH)CO₂H·2H₂O MW: 254.22 g/mol Melting Pt: 105…110 °C |
MDL Number:
MFCD00149540 CAS Number: 5965-83-3 EINECS: 202-555-6 UN: 2585 ADR: 8,III |
Specification Test Results
Assay | 98% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...