Tetramethylammonium hydroxide pentahydrate ≥98%
Supplier: BeanTown Chemical
211480-100G
211480-25G
BT211480-100GEA
156.27
USD
BT211480-100G
BT211480-25G
Tetramethylammonium hydroxide pentahydrate ≥98%
Tetramethylammonium hydroxide pentahydrate
CAS: 10424-65-4; EC No: 200-882-9; MDL No: MFCD00149566; RTECS: PA0875000
UN No: UN3423; Haz Class: 8; Packing Group: II
Crystalline; Linear Formula: (CH3)4NOH·5H2O; Molecular Formula: C4H13NO·5H2O; MW: 181.23
Melting Point: 67-70°
Air Sensitive , Hygroscopic
- RTECS: PA0875000
- Air Sensitive, Hygroscopic
- Crystalline
Formula:
C₄H₁₃NO·5H₂O Melting Pt: 67…70 °C |
MDL Number:
MFCD00149566 CAS Number: 10424-65-4 EINECS: 200-882-9 UN: 3423 ADR: 8,II |
Specification Test Results
Assay | 98% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...