Dibenzothiophene ≥98%
Supplier: BeanTown Chemical
Synonyms:
Dibenzo[b,d]thiophene
211425-100G
211425-25G
211425-500G
BT211425-100GEA
177.17
USD
BT211425-100G
BT211425-25G
BT211425-500G
Dibenzothiophene ≥98%
Dibenzothiophene
CAS: 132-65-0; EC No: 205-072-9; MDL No: MFCD00004969; RTECS: HQ3490550
UN No: UN3077; Haz Class: 9; Packing Group: III
Solid; Molecular Formula: C12H8S; MW: 184.25
Melting Point: 97-100°; Boiling Point: 332-333°; Flash point: 170°C (338°F)
- RTECS: HQ3490550
- Solid
Formula:
C₁₂H₈S MW: 184.26 g/mol Boiling Pt: 332…333 °C Melting Pt: 97…100 °C Flash Pt: 170 °C (338 °F) |
MDL Number:
MFCD00004969 CAS Number: 132-65-0 EINECS: 205-072-9 ADR: 9,III |
Specification Test Results
Assay | 98% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...