N,N-Dimethyloctylamine ≥95%
Supplier: BeanTown Chemical
Synonyms:
N,N-Dimethyloctan-1-amine
211390-10G
211390-250G
211390-50G
BT211390-10GEA
37.55
USD
BT211390-10G
BT211390-250G
BT211390-50G
N,N-Dimethyloctylamine ≥95%
N,N-Dimethyloctylamine
CAS: 7378-99-6; EC No: 230-939-3; MDL No: MFCD00009558
UN No: UN2922; Haz Class: 8 (6.1); Packing Group: III
Liquid; Linear Formula: CH3(CH2)7N(CH3)2; Molecular Formula: C10H23N; MW: 157.30
Melting Point: -57°; Boiling Point: 195°; Flash point: 64°C (147°F)
Density (g/mL): 0.765; Refractive Index: 1.424
Air Sensitive
- Refractive index: 1.424
- Air Sensitive
- Liquid
Formula:
C₁₀H₂₃N MW: 157.3 g/mol Boiling Pt: 195 °C Melting Pt: –57 °C Density: 0.765 g/mL Flash Pt: 64 °C (147 °F) |
MDL Number:
MFCD00009558 CAS Number: 7378-99-6 EINECS: 230-939-3 UN: 2922 ADR: 8,III |
Specification Test Results
Assay | 95% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...