Tungsten(IV) carbide ≥99%

Supplier: BeanTown Chemical

Synonyms: Tungsten monocarbide, Wolfram carbide, Tungsten carbide

211320-250G 211320-50G
BT211320-250GEA 161 USD
BT211320-250G BT211320-50G
Tungsten(IV) carbide ≥99%
Tungsten(IV) carbide

CAS: 12070-12-1; EC No: 235-123-0; MDL No: MFCD00011464; RTECS: YO7250000 UN No: UN3178; Haz Class: 4.1; Packing Group: III -100+270 Mesh Powder; Linear Formula: WC; MW: 195.86 Melting Point: 2785°; Boiling Point: 6000° Density (g/mL): 15.63


  • RTECS: YO7250000
  • -100+270 Mesh Powder

Formula: CW
MW: 195.85 g/mol
Boiling Pt: 6000 °C
Melting Pt: 2785 °C
Density: 15.63 g/mL
MDL Number: MFCD00011464
CAS Number: 12070-12-1
EINECS: 235-123-0
UN: 3178
ADR: 4.1,III

Order Now

Specification Test Results

Assay 99%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR