Isopropyl acetate ≥99%

Supplier: BeanTown Chemical

Synonyms: Acetic acid isopropyl ester

211275-1L 211275-2.5L 211275-250ML
BT211275-1LEA 66.74 USD
BT211275-1L BT211275-2.5L BT211275-250ML
Isopropyl acetate ≥99%
Isopropyl acetate

CAS: 108-21-4; EC No: 203-561-1; MDL No: MFCD00008877; RTECS: AI4930000 UN No: UN1220; Haz Class: 3; Packing Group: II Liquid; Linear Formula: CH3CO2CH(CH3)2; Molecular Formula: C5H10O2; MW: 102.13 Melting Point: -73°; Boiling Point: 85-91°; Flash point: 4°C (39°F) Density (g/mL): 0.872; Refractive Index: 1.377


  • RTECS: AI4930000
  • Refractive index: 1.377
  • Liquid

Formula: C₅H₁₀O₂
MW: 102.13 g/mol
Boiling Pt: 85…91 °C
Melting Pt: –73 °C
Density: 0.872 g/mL
Flash Pt: 4 °C (39 °F)
MDL Number: MFCD00008877
CAS Number: 108-21-4
EINECS: 203-561-1
UN: 1220
ADR: 3,II

Order Now

Specification Test Results

Assay 99%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR