Chromium(III) chloride, anhydrous ≥98%

Supplier: BeanTown Chemical

Synonyms: Chromium trichloride

211255-250G 211255-50G
BT211255-250GEA 301 USD
BT211255-250G BT211255-50G
Chromium(III) chloride, anhydrous ≥98%
Chromium(III) chloride

CAS: 10025-73-7; EC No: 233-038-3; MDL No: MFCD00010948 UN No: UN3260; Haz Class: 8; Packing Group: III Crystalline; Linear Formula: CrCl3; MW: 158.35 Density (g/mL): 2.87 Hygroscopic


  • Hygroscopic
  • Crystalline

Formula: Cl₃Cr
MW: 158.35 g/mol
Melting Pt: 1152
Density: 2.87 g/mL
MDL Number: MFCD00010948
CAS Number: 10025-73-7
EINECS: 233-038-3
UN: 3260
ADR: 8 III
Merck Index: 12,02278

Order Now

Specification Test Results

Assay 98%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR