Calcium perchlorate hydrate
Supplier: BeanTown Chemical
211245-1KG
211245-250G
BT211245-1KGEA
136
USD
BT211245-1KG
BT211245-250G
Calcium perchlorate hydrate
Calcium perchlorate hydrate
CAS: 13477-36-6; EC No: 236-768-0; MDL No: MFCD00015971
UN No: UN1455; Haz Class: 5.1; Packing Group: II
Crystalline; Linear Formula: Ca(ClO4)2·xH2O; MW: 238.98 (anhydrous)
Hygroscopic
- Hygroscopic
- Crystalline
Formula:
Ca(ClO₄)₂·xH₂O MW: 238.98 g/mol (anhy) |
MDL Number:
MFCD00015971 CAS Number: 13477-36-6 EINECS: 236-768-0 UN: 1455 ADR: 5.1 II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...