Cobalt(II) tetrafluoroborate hydrate ≥95%

Supplier: BeanTown Chemical

211235-100G 211235-25G
BT211235-100GEA 97.36 USD
BT211235-100G BT211235-25G
Cobalt(II) tetrafluoroborate hydrate ≥95%
Cobalt(II) tetrafluoroborate hydrate

CAS: 26490-63-1; MDL No: MFCD00016012 UN No: UN3260; Haz Class: 8; Packing Group: III Crystalline; Linear Formula: Co(BF4)2·xH2O; MW: 232.54 (anhydrous) Hygroscopic


  • Hygroscopic
  • Crystalline

Formula: Co(BF₄)₂·xH₂O
MW: 232.54 g/mol (anhy)
MDL Number: MFCD00016012
CAS Number: 26490-63-1
ADR: 8,III

Order Now

Specification Test Results

Assay 95%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR