Rubidium fluoride ≥99.5% (trace metals basis)
Supplier: BeanTown Chemical
211200-10G
211200-50G
BT211200-10GEA
59.48
USD
BT211200-10G
BT211200-50G
Rubidium fluoride ≥99.5% (trace metals basis)
Rubidium fluoride
CAS: 13446-74-7; EC No: 236-603-2; MDL No: MFCD00011191; RTECS: VL8740000
UN No: UN3288; Haz Class: 6.1; Packing Group: III
Pieces/Powder; Linear Formula: RbF; MW: 104.47
Melting Point: 795°
Density (g/mL): 3.557; Refractive Index: 1.398
- RTECS: VL8740000
- Refractive index: 1.398
- Pieces/Powder
Formula:
FRb MW: 104.47 g/mol Melting Pt: 795 °C Density: 3.557 g/mL |
MDL Number:
MFCD00011191 CAS Number: 13446-74-7 EINECS: 236-603-2 UN: 3288 ADR: 6.1,III |
Specification Test Results
Assay | 99.5% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...