4-Chlorobenzonitrile ≥99%

Supplier: BeanTown Chemical

211190-1KG 211190-250G 211190-50G
BT211190-1KGEA 823 USD
BT211190-1KG BT211190-250G BT211190-50G
4-Chlorobenzonitrile ≥99%
4-Chlorobenzonitrile

CAS: 623-03-0; EC No: 210-765-4; MDL No: MFCD00001813; RTECS: DI2800000 UN No: UN3439; Haz Class: 6.1; Packing Group: III Powder/Chunks; Molecular Formula: C7H4ClN; MW: 137.57 Melting Point: 90-93°; Boiling Point: 223°


  • RTECS: DI2800000
  • Powder/Chunks

Formula: C₇H₄ClN
Boiling Pt: 223 °C
Melting Pt: 90…93 °C
MDL Number: MFCD00001813
CAS Number: 623-03-0
EINECS: 210-765-4
ADR: 6.1,III

Order Now

Specification Test Results

Assay 99%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR