Iron(III) isopropoxide 2.5% (w/v) in 2-propanol
Supplier: BeanTown Chemical
Synonyms:
Iron(III) i-propoxide
211175-25ML
211175-5ML
BT211175-25MLEA
166
USD
BT211175-25ML
BT211175-5ML
Iron(III) isopropoxide 2.5% (w/v) in 2-propanol
Iron(III) isopropoxide
CAS: 14995-22-3; MDL No: MFCD00070436
UN No: UN1219; Haz Class: 3; Packing Group: II
Liquid; Linear Formula: Fe[OCH(CH3)2]3 ; Molecular Formula: C9H21FeO3 ; MW: 233.11
Boiling Point: 82°; Flash point: 12°C (54°F)
Density (g/mL): 0.79
Moisture Sensitive
- Moisture Sensitive
- Liquid
Formula:
C₉H₂₁FeO₃ MW: 233.109 g/mol Boiling Pt: 82 °C Density: 0.79 g/mL Flash Pt: 12 °C (54 °F) |
MDL Number:
MFCD00070436 CAS Number: 14995-22-3 ADR: 3,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...