Disodium tetrasulphide ≥90%, Technical Grade
Supplier: BeanTown Chemical
Synonyms:
Sodium tetrasulphide, Sodium sulphide
211150-10G
211150-50G
BT211150-10GEA
122
USD
BT211150-10G
BT211150-50G
Disodium tetrasulphide ≥90%, Technical Grade
Disodium tetrasulphide
CAS: 12034-39-8; EC No: 215-211-5; MDL No: MFCD00014249
UN No: UN1385; Haz Class: 4.2; Packing Group: II
Powder; Molecular Formula: Na2S4 ; MW: 174.24
Melting Point: 275°
Hygroscopic, Moisture Sensitive
- Hygroscopic, Moisture Sensitive
- Powder
Formula:
Na₂S₄ Melting Pt: 275 °C |
MDL Number:
MFCD00014249 CAS Number: 12034-39-8 EINECS: 215-211-5 UN: 1385 ADR: 4.2,II |
Specification Test Results
Assay | 90% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...