Zinc fluoride tetrahydrate ≥98%
Supplier: BeanTown Chemical
211140-100G
211140-500G
BT211140-100GEA
49
USD
BT211140-100G
BT211140-500G
Zinc fluoride tetrahydrate ≥98%
Zinc fluoride tetrahydrate
CAS: 13986-18-0; EC No: 232-001-9; MDL No: MFCD00149892
UN No: UN3288; Haz Class: 6.1; Packing Group: III
Crystalline; Linear Formula: ZnF2·4H2O; MW: 175.45
Melting Point: 872-910°
- Crystalline
Formula:
F₂Zn·4H₂O MW: 175.45 Melting Pt: 872 Density: 2.255 |
MDL Number:
MFCD00149892 CAS Number: 13986-18-0 EINECS: 232-001-9 UN: 3288 ADR: 6.1 III |
Specification Test Results
Assay | 98% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...