Lead(II) chromate ≥98.0% ACS
Supplier: BeanTown Chemical
211075-100G
211075-500G
BT211075-100GEA
62.64
USD
BT211075-100G
BT211075-500G
Lead(II) chromate ≥98.0% ACS
CAS: 7758-97-6; EC No: 231-846-0; MDL No: MFCD00011158; RTECS: GB2975000
UN No: UN3077; Haz Class: 9; Packing Group: III
Powder; Linear Formula: PbCrO4; MW: 323.19
- RTECS: GB2975000
- Powder
Formula:
CrO₄Pb |
MDL Number:
MFCD00011158 CAS Number: 7758-97-6 EINECS: 231-846-0 UN: 3077 ADR: 6.1,III |
Specification Test Results
Assay | 98.0% min. |
Soluble matter | ≤0.15% |
Carbon compounds (as C) | ≤0.01% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...