4-tert-Butylbenzoyl chloride ≥98%

Supplier: BeanTown Chemical

Synonyms: 4-tert-Butylbenzoylchloride

211070-100G 211070-25G 211070-500G
BT211070-100GEA 110 USD
BT211070-100G BT211070-25G BT211070-500G
4-tert-Butylbenzoyl chloride ≥98%
4-tert-Butylbenzoyl chloride

CAS: 1710-98-1; EC No: 216-973-1; MDL No: MFCD00000695 UN No: UN3265; Haz Class: 8; Packing Group: II Liquid; Molecular Formula: C11H13ClO; MW: 196.67 Boiling Point: 135°/20 mmHg; Flash point: 87°C (189°F) Density (g/mL): 1.007; Refractive Index: 1.536 Moisture Sensitive


  • Refractive index: 1.536
  • Moisture Sensitive
  • Liquid

Formula: C₁₁H₁₃ClO
MW: 196.68 g/mol
Boiling Pt: 135 °C/20 mmHg
Density: 1.007 g/mL
Flash Pt: 87 °C (189 °F)
MDL Number: MFCD00000695
CAS Number: 1710-98-1
EINECS: 216-973-1
UN: 3265
ADR: 8,II

Order Now

Specification Test Results

Assay 98%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR