5-Bromo-1-pentene ≥96%

Supplier: BeanTown Chemical

211005-1G 211005-25G 211005-5G
BT211005-1GEA 22 USD
BT211005-1G BT211005-25G BT211005-5G
5-Bromo-1-pentene ≥96%
5-Bromo-1-pentene

CAS: 1119-51-3; EC No: 214-281-4; MDL No: MFCD0000-364 UN No: UN1993; Haz Class: 3; Packing Group: III Liquid; Linear Formula: H2C=CH(CH2)3Br; Molecular Formula: C5H9Br; MW: 149.03 Boiling Point: 126-127°; Flash point: 31°C (88°F) Density (g/mL): 1.258; Refractive Index: 1.463


  • Refractive index: 1.463
  • Liquid

Formula: C₅H₉Br
MW: 149.03 g/mol
Boiling Pt: 126…127 °C
Density: 1.258 g/mL
Flash Pt: 31 °C (88 °F)
MDL Number: MFCD00000264
CAS Number: 1119-51-3
EINECS: 214-281-4
UN: 1993
ADR: 3,III

Order Now

Specification Test Results

Assay 96%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR