Adenine ≥99%

Supplier: BeanTown Chemical

Synonyms: Purin-6-amine, 7H-Purin-6-amine, 6-Aminopurine

145965-250G 145965-50G
BT145965-250GEA 428 USD
BT145965-250G BT145965-50G
Adenine ≥99%
Adenine

CAS: 73-24-5; EC No: 200-796-1; MDL No: MFCD00041790; RTECS: AU6125000 UN No: UN2811; Haz Class: 6.1; Packing Group: III Powder; Molecular Formula: C5H5N5; MW: 135.13 Melting Point: 360°


  • RTECS: AU6125000
  • Powder

Formula: C₅H₅N₅
MW: 135.13 g/mol
Melting Pt: >360 °C
MDL Number: MFCD00041790
CAS Number: 73-24-5
EINECS: 200-796-1
UN: 2811
ADR: 6.1 III
Merck Index: 13,00152

Order Now

Specification Test Results

Assay 99%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR