Barium tetracyanoplatinate(II) tetrahydrate ≥99.99% (trace metals basis)
Supplier: BeanTown Chemical
145865-1G
145865-5G
BT145865-1GEA
301
USD
BT145865-1G
BT145865-5G
Barium tetracyanoplatinate(II) tetrahydrate ≥99.99% (trace metals basis)
Barium tetracyanoplatinate(II) tetrahydrate
CAS: 13755-32-3; EC No: 209-238-1; MDL No: MFCD00014183
UN No: UN1564; Haz Class: 6.1; Packing Group: II
Powder; Linear Formula: BaPt(CN)4·4H2O; MW: 508.56
- Powder
Formula:
BaPt(CN)₄.4H₂O |
MDL Number:
MFCD00014183 CAS Number: 13755-32-3 EINECS: 209-238-1 ADR: 6.1,II |
Specification Test Results
Assay | 99.99% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...