n-Octane ≥98%

Supplier: BeanTown Chemical

145860-100ML 145860-2.5L 145860-500ML
BT145860-100MLEA 37.48 USD
BT145860-100ML BT145860-2.5L BT145860-500ML
n-Octane ≥98%
n-Octane

CAS: 111-65-9; EC No: 203-892-1; MDL No: MFCD00009556; RTECS: RG8400000 UN No: UN1262; Haz Class: 3; Packing Group: II Liquid; Linear Formula: CH3(CH2)6CH3; Molecular Formula: C8H18; MW: 114.23 Melting Point: -57°; Boiling Point: 125-127°; Flash point: 12°C (55°F) Density (g/mL): 0.703; Refractive Index: 1.398


  • RTECS: RG8400000
  • Refractive index: 1.398
  • Liquid

Formula: H₃C(CH₂)₆CH₃
MW: 114.23 g/mol
Boiling Pt: 125…127 °C
Melting Pt: –57 °C
Density: 0.703 g/mL
Flash Pt: 12 °C (55 °F)
MDL Number: MFCD00009556
CAS Number: 111-65-9
EINECS: 203-892-1
UN: 1262
ADR: 3,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=68adf890d68728825d4a2fa4b032700b93f710b8&vid=b00a2925902bc425c80f8ebb3f0e7b456c800ae3&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay 98%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR