4-Octyne ≥99%
Supplier: BeanTown Chemical
Synonyms:
Dipropylacetylene
145830-25G
145830-5G
BT145830-25GEA
79
USD
BT145830-25G
BT145830-5G
4-Octyne ≥99%
4-Octyne
CAS: 1942-45-6; EC No: 217-730-2; MDL No: MFCD00009471
UN No: UN3295; Haz Class: 3; Packing Group: II
Liquid; Linear Formula: CH3CH2CH2C≡CCH2CH2CH3; Molecular Formula: C8H14; MW: 110.20
Melting Point: -103°; Boiling Point: 131-132°; Flash point: 29°C (84°F)
Density (g/mL): 0.751; Refractive Index: 1.425
- Refractive index: 1.425
- Liquid
Formula:
C₈H₁₄ MW: 110.2 g/mol Boiling Pt: 131…132 °C Melting Pt: –103 °C Density: 0.751 g/mL Flash Pt: 29 °C (84 °F) |
MDL Number:
MFCD00009471 CAS Number: 1942-45-6 EINECS: 217-730-2 UN: 3295 ADR: 3,II |
Specification Test Results
Assay | 99% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...