Potassium fluoride ≥99.99%
Supplier: BeanTown Chemical
145825-1G
145825-5G
BT145825-1GEA
14
USD
BT145825-1G
BT145825-5G
Potassium fluoride ≥99.99%
Potassium fluoride
CAS: 7789-23-3; EC No: 232-151-5; MDL No: MFCD00011398; RTECS: TT0700000
UN No: UN1812; Haz Class: 6.1; Packing Group: III
Powder/Chunks; Linear Formula: KF; MW: 58.1
Melting Point: 858°; Boiling Point: 1505°
Density (g/mL): 2.48; Refractive Index: 1.363
Hygroscopic
- RTECS: TT0700000
- Refractive index: 1.363
- Hygroscopic
- Powder/Chunks
Formula:
KF MW: 58.1 g/mol Boiling Pt: 1505 Melting Pt: 860 Density: 2.48 g/mL |
MDL Number:
MFCD00011398 CAS Number: 7789-23-3 EINECS: 232-151-5 UN: 1812 ADR: 6.1 III Merck Index: 13,07714 |
Specification Test Results
Assay | 99.99% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...