Diethylenetriamine ≥99%
Supplier: BeanTown Chemical
Synonyms:
2,2'-Iminodi(ethylamine), DIEN, Diethylenetriamine
145800-100ML
145800-2.5L
145800-500ML
BT145800-100MLEA
32.84
USD
BT145800-100ML
BT145800-2.5L
BT145800-500ML
Diethylenetriamine ≥99%
Diethylenetriamine
CAS: 111-40-0; EC No: 203-865-4; MDL No: MFCD00008171; RTECS: IE1225000
UN No: UN2079; Haz Class: 8; Packing Group: II
Liquid; Linear Formula: (NH2CH2CH2)2NH; Molecular Formula: C4H13N3; MW: 103.17
Melting Point: -35°; Boiling Point: 199-209°; Flash point: 102°C (216°F)
Density (g/mL): 0.955; Refractive Index: 1.483
Air Sensitive
- RTECS: IE1225000
- Refractive index: 1.483
- Air Sensitive
- Liquid
Formula:
C₄H₁₃N₃ Boiling Pt: 199…209 °C Melting Pt: –35 °C Density: 0.955 g/mL Flash Pt: 102 °C (216 °F) |
MDL Number:
MFCD00008171 CAS Number: 111-40-0 EINECS: 203-865-4 UN: 2079 ADR: 8,II |
Specification Test Results
Assay | 99% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...