Cyclohexylbenzene ≥97%

Supplier: BeanTown Chemical

Synonyms: Phenylcyclohexane

145740-100G 145740-500G
BT145740-100GEA 46 USD
BT145740-100G BT145740-500G
Cyclohexylbenzene ≥97%
Cyclohexylbenzene

CAS: 827-52-1; EC No: 212-572-0; MDL No: MFCD00001451; RTECS: CZ1330000 UN No: UN3082; Haz Class: 9; Packing Group: III Liquid; Molecular Formula: C12H16; MW: 160.26 Melting Point: 4-7°; Boiling Point: 239-240°; Flash point: 81°C (178°F) Density (g/mL): 0.95; Refractive Index: 1.526


  • RTECS: CZ1330000
  • Refractive index: 1.526
  • Liquid

Formula: C₁₂H₁₆
MW: 160.26 g/mol
Boiling Pt: 239…240 °C
Melting Pt: 4…7 °C
Density: 0.95 g/mL
Flash Pt: 81 °C (178 °F)
MDL Number: MFCD00001451
CAS Number: 827-52-1
EINECS: 212-572-0
UN: 3082
ADR: 9,III

Order Now

Specification Test Results

Assay 97%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR