4,6-Dichloropyrimidine ≥98%
Supplier: BeanTown Chemical
145735-25G
145735-5G
BT145735-25GEA
150
USD
BT145735-25G
BT145735-5G
4,6-Dichloropyrimidine ≥98%
4,6-Dichloropyrimidine
CAS: 1193-21-1; EC No: 214-770-2; MDL No: MFCD00006109
UN No: UN3261; Haz Class: 8; Packing Group: II
Crystalline; Molecular Formula: C4H2Cl2N2; MW: 148.98
Melting Point: 65-67°; Boiling Point: 176°
- Crystalline
Formula:
C₄H₂Cl₂N₂ MW: 148.98 g/mol Boiling Pt: 176 °C Melting Pt: 65…67 °C |
MDL Number:
MFCD00006109 CAS Number: 1193-21-1 EINECS: 214-770-2 UN: 3261 ADR: 8,II |
Specification Test Results
Assay | 98% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...