Propyl acetate ≥99%
Supplier: BeanTown Chemical
Synonyms:
Acetic acid propyl ester
145685-1L
145685-500ML
BT145685-1LEA
36
USD
BT145685-1L
BT145685-500ML
Propyl acetate ≥99%
Propyl acetate
CAS: 109-60-4; EC No: 203-686-1; MDL No: MFCD00009372; RTECS: AJ3675000
UN No: UN1276; Haz Class: 3; Packing Group: II
Liquid; Linear Formula: CH3COOCH2CH2CH3; Molecular Formula: C5H10O2; MW: 102.13
Melting Point: -95°; Boiling Point: 102°; Flash point: 14°C (57°F)
Density (g/mL): 0.888; Refractive Index: 1.384
- RTECS: AJ3675000
- Refractive index: 1.384
- Liquid
Formula:
C₅H₁₀O₂ Boiling Pt: 102 °C Melting Pt: –95 °C Density: 0.888 g/mL Flash Pt: 14 °C (57 °F) |
MDL Number:
MFCD00009372 CAS Number: 109-60-4 EINECS: 203-686-1 UN: 1276 ADR: 3,II |
Specification Test Results
Assay | 99% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...