Trioctylamine ≥95%

Supplier: BeanTown Chemical

Synonyms: Tri-n-octylamine

145610-100ML 145610-2.5L 145610-500ML
BT145610-100MLEA 58 USD
BT145610-100ML BT145610-2.5L BT145610-500ML
Trioctylamine ≥95%
Trioctylamine

CAS: 1116-76-3; EC No: 214-242-1; MDL No: MFCD00009560; RTECS: RG8225000 UN No: UN3082; Haz Class: 9; Packing Group: III Liquid; Linear Formula: [CH3(CH2)7]3N; Molecular Formula: C24H51N; MW: 353.68 Melting Point: -39°; Flash point: 158°C (316°F) Density (g/mL): 0.809; Refractive Index: 1.449 Air Sensitive


  • RTECS: RG8225000
  • Refractive index: 1.449
  • Air Sensitive
  • Liquid

Formula: C₂₄H₅₁N
MW: 353.68 g/mol
Melting Pt: –39 °C
Density: 0.809 g/mL
Flash Pt: 158 °C (316 °F)
MDL Number: MFCD00009560
CAS Number: 1116-76-3
EINECS: 214-242-1
UN: 3082
ADR: 9,III

Order Now

Specification Test Results

Assay 95%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR