Magnesium nitride ≥99.5% (trace metals basis)
Supplier: BeanTown Chemical
Synonyms:
Trimagnesium dinitride
145555-10G
145555-50G
BT145555-10GEA
79.68
USD
BT145555-10G
BT145555-50G
Magnesium nitride ≥99.5% (trace metals basis)
Magnesium nitride
CAS: 12057-71-5; EC No: 235-022-1; MDL No: MFCD00016205
UN No: UN3178; Haz Class: 4.1; Packing Group: II
-325 Mesh Powder; Molecular Formula: Mg3N2 ; MW: 100.95
Melting Point: 800° (decomposes)
Density (g/mL): 2.71
Air Sensitive, Moisture Sensitive
- Air Sensitive, Moisture Sensitive
- -325 Mesh Powder
Formula:
Mg₃N₂ MW: 100.93 g/mol Melting Pt: 800 °C (dec.) Density: 2.71 g/mL |
MDL Number:
MFCD00016205 CAS Number: 12057-71-5 EINECS: 235-022-1 UN: 3178 ADR: 4.1,II |
Specification Test Results
Assay | 99.5% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...