1,3,5-Trichlorobenzene ≥98%

Supplier: BeanTown Chemical

145535-250G 145535-50G
BT145535-250GEA 107 USD
BT145535-250G BT145535-50G
1,3,5-Trichlorobenzene ≥98%
1,3,5-Trichlorobenzene

CAS: 108-70-3; EC No: 203-608-6; MDL No: MFCD00000585; RTECS: DC2100100 UN No: UN3077; Haz Class: 9; Packing Group: III Crystalline; Molecular Formula: C6H3Cl3 ; MW: 181.45 Melting Point: 56-60°; Boiling Point: 208°; Flash point: 107°C (225°F)


  • RTECS: DC2100100
  • Crystalline

Formula: C₆H₃Cl₃
MW: 181.45 g/mol
Boiling Pt: 208 °C
Melting Pt: 56…60 °C
Flash Pt: 107 °C (225 °F)
MDL Number: MFCD00000585
CAS Number: 108-70-3
EINECS: 203-608-6
ADR: 9,III

Order Now

Specification Test Results

Assay 98%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR