Thiomorpholine ≥98%
Supplier: BeanTown Chemical
145475-100G
145475-25G
145475-5G
BT145475-100GEA
625
USD
BT145475-100G
BT145475-25G
BT145475-5G
Thiomorpholine ≥98%
Thiomorpholine
CAS: 123-90-0; EC No: 204-660-2; MDL No: MFCD00005974
UN No: UN3267; Haz Class: 8; Packing Group: III
Liquid; Molecular Formula: C4H9NS; MW: 103.19
Boiling Point: 169°; Flash point: 63°C (145°F)
Density (g/mL): 1.026; Refractive Index: 1.538
Air Sensitive
- Refractive index: 1.538
- Air Sensitive
- Liquid
Formula:
C₄H₉NS MW: 103.19 g/mol Boiling Pt: 169 °C Density: 1.026 g/mL Flash Pt: 63 °C (145 °F) |
MDL Number:
MFCD00005974 CAS Number: 123-90-0 EINECS: 204-660-2 UN: 3267 ADR: 8,III |
Specification Test Results
Assay | 98% |
Learn more
![](/m-us.vwr.com/en_US/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...