Hydriodic acid ≥47.0% stabilized ACS
Supplier: BeanTown Chemical
Synonyms:
Hydroiodic acid, Hydrogen iodide, Hydrogen iodide solution
145405-100ML
145405-500ML
BT145405-100MLEA
80.61
USD
BT145405-100ML
BT145405-500ML
Hydriodic acid ≥47.0% stabilized ACS
Hydriodic acid
CAS: 10034-85-2; EC No: 233-109-9; MDL No: MFCD00011347; RTECS: MW3760000
UN No: UN1787; Haz Class: 8; Packing Group: II
Liquid; Molecular Formula: HI; MW: 127.91
Boiling Point: 127°
Density (g/mL): 1.701
Light Sensitive
- RTECS: MW3760000
- Light Sensitive
- Liquid
Formula:
HI MW: 127.91 g/mol Boiling Pt: 127 °C Density: 1.701 g/mL |
MDL Number:
MFCD00011347 CAS Number: 10034-85-2 EINECS: 233-109-9 UN: 1787 ADR: 8,II |
Specification Test Results
Assay | 47.0% min. |
Chloride & bromide | ≤0.05% |
Sulfate (SO4) | ≤0.005% |
Iron (Fe) | ≤0.001% |
Stabilizer (H3PO2) | 1.5% |
Heavy metals (as Pb) | ≤0.001% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...