O-Methylhydroxylamine hydrochloride ≥98%
Supplier: BeanTown Chemical
Synonyms:
Methylhydroxylamine hydrochloride, O-Methylhydroxylammonium chloride, Methoxyamine hydrochloride, Methoxylamine hydrochloride
145395-25G
145395-5G
BT145395-25GEA
197
USD
BT145395-25G
BT145395-5G
O-Methylhydroxylamine hydrochloride ≥98%
O-Methylhydroxylamine hydrochloride
CAS: 593-56-6; EC No: 209-798-7; MDL No: MFCD00012951; RTECS: NC3980000
UN No: UN3261; Haz Class: 8; Packing Group: III
Crystalline; Linear Formula: CH3ONH2·HCl; MW: 83.52
Melting Point: 151-154°
Hygroscopic
- RTECS: NC3980000
- Hygroscopic
- Crystalline
Formula:
CH₆ClNO MW: 83.515 g/mol Melting Pt: 151…154 °C |
MDL Number:
MFCD00012951 CAS Number: 593-56-6 EINECS: 209-798-7 UN: 3261 ADR: 8,III |
Specification Test Results
Assay | 98% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...