2-(Chloromethyl)-4-methoxy-3,5-dimethylpyridine hydrochloride ≥98%
Supplier: BeanTown Chemical
Synonyms:
2-Chloromethyl-4-methoxy-3,5-dimethylpyridine hydrochloride
145365-25G
145365-5G
BT145365-25GEA
155
USD
BT145365-25G
BT145365-5G
2-(Chloromethyl)-4-methoxy-3,5-dimethylpyridine hydrochloride ≥98%
2-(Chloromethyl)-4-methoxy-3,5-dimethylpyridine hydrochloride
CAS: 86604-75-3; MDL No: MFCD00277472
UN No: UN3261; Haz Class: 8; Packing Group: II
Solid; Molecular Formula: C9H12ClNO·HCl ; MW: 222.11
Melting Point: 128-131°
- Solid
Formula:
C₉H₁₂ClNO MW: 222.11 g/mol Melting Pt: 128…131 °C |
MDL Number:
MFCD00277472 CAS Number: 86604-75-3 ADR: 8,II |
Specification Test Results
Assay | 98% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...