Mercury(II) perchlorate trihydrate
Supplier: BeanTown Chemical
145360-100G
145360-25G
145360-500G
BT145360-100GEA
154.57
USD
BT145360-100G
BT145360-25G
BT145360-500G
Mercury(II) perchlorate trihydrate
Mercury(II) perchlorate trihydrate
CAS: 73491-34-6; EC No: 231-525-5; MDL No: MFCD00149735
UN No: UN3087; Haz Class: 5.1 (6.1); Packing Group: II
Crystalline; Linear Formula: Hg(ClO4)2·3H2O; MW: 453.54
- Crystalline
Formula:
Cl₂HgO₈·3H₂O |
MDL Number:
MFCD00149735 CAS Number: 73491-34-6 EINECS: 231-525-5 UN: 3087 ADR: 5.1,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...