Tantalum ≥99.9% (trace metals basis), Powder -325 mesh

Supplier: BeanTown Chemical

145355-100G 145355-25G
BT145355-100GEA 396.49 USD
BT145355-100G BT145355-25G
Tantalum ≥99.9% (trace metals basis), Powder -325 mesh
Tantalum

CAS: 7440-25-7; EC No: 231-135-5; MDL No: MFCD00011252; RTECS: WW5505000 UN No: UN3089; Haz Class: 4.1; Packing Group: II Powder; Molecular Formula: Ta; MW: 180.95 Melting Point: 2996°; Boiling Point: 5425° Density (g/mL): 16.69


  • RTECS: WW5505000
  • Powder

Formula: Ta
MW: 180.95 g/mol
Boiling Pt: 5425 °C
Melting Pt: 2996 °C
Density: 16.69 g/mL
MDL Number: MFCD00011252
CAS Number: 7440-25-7
EINECS: 231-135-5
UN: 3089
ADR: 4.1,II

Order Now

Specification Test Results

Assay 99.9%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR