Spermine-(N,N'-bis(3-aminopropyl)-1,4-butanediamine) ≥97%

Supplier: BeanTown Chemical

Synonyms: N1,N1'-(Butane-1,4-diyl)bis(propane-1,3-diamine), Spermine

145330-1G 145330-5G
BT145330-1GEA 54.87 USD
BT145330-1G BT145330-5G
Spermine-(N,N'-bis(3-aminopropyl)-1,4-butanediamine) ≥97%
Spermine-(N,N'-bis(3-aminopropyl)-1,4-butanediamine)

CAS: 71-44-3; EC No: 200-754-2; MDL No: MFCD00008215; RTECS: EJ7230000 UN No: UN3259 ; Haz Class: 8; Packing Group: III Solid; Linear Formula: NH2(CH2)3NH(CH2)4NH(CH2)3NH2; Molecular Formula: C10H26N4; MW: 202.34 Melting Point: 28-30°; Boiling Point: 150°/5 mmHg; Flash point: 110°C (230°F) Density (g/mL): 0.937 Air Sensitive , Hygroscopic


  • RTECS: EJ7230000
  • Air Sensitive, Hygroscopic
  • Solid

Formula: C₁₀H₂₆N₄
MW: 202.35 g/mol
Boiling Pt: 150 °C/5 mmHg
Melting Pt: 28…30 °C
Density: 0.937 g/mL
Flash Pt: 110 °C (230 °F)
MDL Number: MFCD00008215
CAS Number: 71-44-3
EINECS: 200-754-2
ADR: 8,III

Order Now

Specification Test Results

Assay 97%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR