Zirconium ≥99.9% (trace rare earth metals basis), powder
Supplier: BeanTown Chemical
145325-100G
145325-25G
145325-5G
BT145325-100GEA
262.9
USD
BT145325-100G
BT145325-25G
BT145325-5G
Zirconium ≥99.9% (trace rare earth metals basis), powder
Zirconium
CAS: 7440-67-7; EC No: 231-176-9; MDL No: MFCD00011303; RTECS: ZH7070000
UN No: UN1358; Haz Class: 4.1; Packing Group: II
Powder, packaged in water; Molecular Formula: Zr; MW: 91.22
Melting Point: 1852°; Boiling Point: 4377°
Density (g/mL): 6.5
- RTECS: ZH7070000
- Powder, packaged in water
Formula:
Zr MW: 91.22 g/mol Boiling Pt: 4377 °C Melting Pt: 1852 °C Density: 6.5 g/mL |
MDL Number:
MFCD00011303 CAS Number: 7440-67-7 EINECS: 231-176-9 UN: 1358 ADR: 4.2 II |
Specification Test Results
Assay | 99.9% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...