2,6-Lutidine ≥98%

Supplier: BeanTown Chemical

Synonyms: 2,6-Dimethylpyridine

145295-100ML 145295-25ML 145295-500ML
BT145295-100MLEA 37.48 USD
BT145295-100ML BT145295-25ML BT145295-500ML
2,6-Lutidine ≥98%
2,6-Lutidine

CAS: 108-48-5; EC No: 203-587-3; MDL No: MFCD00006345; RTECS: OK9700000 UN No: UN1993; Haz Class: 3; Packing Group: III Liquid; Molecular Formula: C7H9N; MW: 107.15 Melting Point: -6°; Boiling Point: 143-145° Density (g/mL): 0.92; Refractive Index: 1.497 Hygroscopic


  • RTECS: OK9700000
  • Refractive index: 1.497
  • Hygroscopic
  • Liquid

Formula: C₇H₉N
MW: 107.16 g/mol
Boiling Pt: 143…145 °C
Melting Pt: –6 °C
Density: 0.92 g/mL
MDL Number: MFCD00006345
CAS Number: 108-48-5
EINECS: 203-587-3
UN: 1993
ADR: 3,III

Order Now

Specification Test Results

Assay 98%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR