5-Amino-2-methylpyridine ≥97%

Supplier: BeanTown Chemical

Synonyms: 5-Amino-2-picoline, 6-Methylpyridin-3-amine, 3­Amino­6­methylpyridine, 6-Methyl-3-pyridinamine, 3­Amino­6­picoline

145290-1G 145290-250MG
BT145290-1GEA 212 USD
BT145290-1G BT145290-250MG
5-Amino-2-methylpyridine ≥97%
5-Amino-2-methylpyridine

CAS: 3430-14-6; MDL No: MFCD00833389 UN No: UN2811; Haz Class: 6.1; Packing Group: III Solid; Molecular Formula: C6H8N2 ; MW: 108.14 Melting Point: 95-99°


  • Solid

Formula: C₆H₈N₂
MW: 108.14
Melting Pt: 95…99 °C
MDL Number: MFCD00833389
CAS Number: 3430-14-6
UN: 2811
ADR: 6.1 III

Order Now

Specification Test Results

Assay 97%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR