Methyl-4-bromocrotonate ≥85%, Technical Grade
Supplier: BeanTown Chemical
145280-10G
145280-250G
145280-50G
BT145280-10GEA
42.14
USD
BT145280-10G
BT145280-250G
BT145280-50G
Methyl-4-bromocrotonate ≥85%, Technical Grade
Methyl-4-bromocrotonate
CAS: 1117-71-1; EC No: 214-251-0; MDL No: MFCD00000246; RTECS: GQ3120000
UN No: UN1760; Haz Class: 8; Packing Group: III
Liquid; Linear Formula: BrCH2CH=CHCOOCH3; Molecular Formula: C5H7BrO2; MW: 179.01
Boiling Point: 83-85°/13 mmHg; Flash point: 91°C (195°F)
Density (g/mL): 1.522; Refractive Index: 1.501
- RTECS: GQ3120000
- Refractive index: 1.501
- Liquid
Formula:
C₅H₇BrO₂ Boiling Pt: 83…85 °C/13 mmHg Density: 1.522 g/mL Flash Pt: 91 °C (195 °F) |
MDL Number:
MFCD00000246 CAS Number: 1117-71-1 EINECS: 214-251-0 ADR: 8,III |
Specification Test Results
Assay | 85% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...