3-Isocyanatopropyltriethoxysilane ≥95%

Supplier: BeanTown Chemical

Synonyms: ICPTES, Triethoxy(3-isocyanatopropyl)silane, (3-Isocyanatopropyl)triethoxysilane, Isocyanic acid 3-(Triethoxysilyl)propyl ester, 3-(Triethoxysilyl)propyl Isocyanate

145235-25G 145235-5G
BT145235-25GEA 173 USD
BT145235-25G BT145235-5G
3-Isocyanatopropyltriethoxysilane ≥95%
3-Isocyanatopropyltriethoxysilane

CAS: 24801-88-5; EC No: 246-467-6; MDL No: MFCD00051459; RTECS: VV6691000 UN No: UN2927; Haz Class: 6.1 (8); Packing Group: II Liquid; Molecular Formula: C10H21NO4Si; MW: 247.36 Boiling Point: 238°; Flash point: 77°C (171°F) Density (g/mL): 0.999; Refractive Index: 1.420 Moisture Sensitive


  • RTECS: VV6691000
  • Refractive index: 1.420
  • Moisture Sensitive
  • Liquid

Formula: C₁₀H₂₁NO₄Si
MW: 247.366 g/mol
Boiling Pt: 238 °C
Density: 0.999 g/mL
Flash Pt: 77 °C (171 °F)
MDL Number: MFCD00051459
CAS Number: 24801-88-5
EINECS: 246-467-6
ADR: 6.1 (8),II

Order Now

Specification Test Results

Assay 95%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR